missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (NIH patent anti-Mesothelin) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28582JF525
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Mesothelin | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| Mesothelin | |
| 0.1 mL | |
| Cancer | |
| 10232 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| NIH patent anti-Mesothelin | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction