missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Nucleoprotein Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33298-20ul
This item is not returnable.
View return policy
Description
MERS-CoV Nucleoprotein Monoclonal antibody specifically detects MERS-CoV Nucleoprotein in MERS-CoV samples. It is validated for ELISA,Western Blot
Specifications
| MERS-CoV Nucleoprotein | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| MERS, MERS NC, MERS Nucleocapsid protein, MERS Nucleoprotein, MERSCoV NC, MERS-CoV NC, MERSCoV Nucleocapsid protein, MERS-CoV Nucleocapsid protein, MERSCOV Nucleoprotein, MERS-CoV Protein N, Middle East respiratory syndrome-related coronavirus, Nucleocapsid protein, Protein N | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-413 of MERS-CoV Nucleoprotein (YP_009047211.1).,, Sequence:, MASPAAPRAVSFADNNDITNTNLSRGRGRNPKPRAAPNNTVSWYTGLTQHGKVPLTFPPGQGVPLNANSTPAQNAGYWRRQDRKINTGNGIKQLAPRWYFYYTGTGPEAALPFRAVKDGIVWVHEDGATDAPSTFGTRNPNNDSAIVTQFAPGTKLPKNFHIEGTGGNSQSSSRASSLSRNSSRSSSQGSRSGNSTRGTSPGPSGIGAVGGDLLYLDLLNRLQALESGKVKQSQPKVITKKDAAAAKNKMRHKRTSTKSFNMVQAFGLRGPGDLQGNFGDLQLNKLGTEDPRWPQIAELAPTASAFMGMSQFKLTHQNNDDHGNPVYFLRYSGAIKLDPKNPNYNKWLELLEQNIDAYKTFPKKEKKQKAPKEESTDQMSEPPKEQRVQGSITQRTRTRPSVQPGPMIDVNTD | |
| 20 μL | |
| Primary | |
| MERS-CoV | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 37616439 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction