missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meprin beta Subunit/MEP1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | Meprin beta Subunit/MEP1B |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630596
|
Novus Biologicals
NBP2-99477-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655505
|
Novus Biologicals
NBP2-99477-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Meprin beta Subunit/MEP1B Polyclonal antibody specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for ELISASpecifications
| Meprin beta Subunit/MEP1B | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Proteases & Other Enzymes | |
| 0.2 um filtered solution in PBS | |
| 4225 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.24.63, endopeptidase-2, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta | |
| Produced in rabbits immunized with purified, recombinant Human Meprin beta Subunit/MEP1B (Accession#: NP_005916.2; Met1-Thr652) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title