Learn More
Novus Biologicals™ Melanotransferrin/CD228/MFI2 Antibody (SC-005) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28837-50ug
This item is not returnable.
View return policy
Description
Melanotransferrin/CD228/MFI2 Monoclonal antibody specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Melanotransferrin/CD228/MFI2 | |
| Monoclonal | |
| Unconjugated | |
| P08582 | |
| Human | |
| Protein A purified | |
| RUO | |
| 4241 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| SC-005 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MGC4856, MTF1 | |
| Melanotransferrin/ CD228 | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |