missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MEKK2 Antibody (SR3035), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00€ - 382.00€
Specifications
| Antigen | MEKK2 |
|---|---|
| Clone | SR3035 |
| Dilution | Western Blot 1:5000-1:20000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041771
|
Novus Biologicals™
NBP3-27740-100ul |
100 μL |
382.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041559
|
Novus Biologicals™
NBP3-27740-50ul |
50 μL |
316.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEKK2 Monoclonal antibody specifically detects MEKK2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| MEKK2 | |
| Western Blot 1:5000-1:20000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11.25, MAP/ERK kinase kinase 2, MAPK/ERK kinase kinase 2, MAPKKK2, MEK kinase 2, MEKK 2, MEKK2, MEKK2B, mitogen-activated protein kinase kinase kinase 2 | |
| A synthesized peptide derived from human MEKK2 (Uniprot #: Q9Y2U5) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3035 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Autophagy, Cancer, Mitophagy, Protein Kinase | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 10746 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title