missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67872-1-IG-150UL
This item is not returnable.
View return policy
Description
The MEK1 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human MEK1. This antibody recognizes human, rat, mouse antigen. The MEK1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| MEK1 | |
| Monoclonal | |
| 1000 μg/mL | |
| MAPKK1; MEK1; MKK1; PRKMK1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Rat, Mouse | |
| 40-50 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 3F6D7 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5604 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction