Learn More
MEIS1 Antibody (OTI3H2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44994-100ul
 View more versions of this product
Product Code. 30306212
  • 550.00€ / 100µL
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MEIS1
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
4211
Store at -20 degrees C. Avoid freeze/thaw cycles.
Purified
Immunohistochemistry, Western Blot
OTI3H2
Western Blot 1:2000, Immunohistochemistry 1:500
homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse)
Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1(NP_002389) produced in E.coli.
100 μL
Primary
Human
Antibody