missing translation for 'onlineSavingsMsg'
Learn More

Medroxyprogesterone Acetate, Mikromol™

Product Code. 18051880
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 18051880

Brand: Mikromol MM0532.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Medroxyprogesterone Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Medroxyprogesterone acetate
CAS 71-58-9
Analyte or Component Names Medroxyprogesterone acetate
Shipping Condition Room Temperature
Molecular Formula C24 H34 O4
Certifications/Compliance ISO 17034
Synonym 6alpha-Methyl-3,20-dioxopregn-4-en-17-yl acetate, Medroxyprogesterone acetate, Megestrol Acetate Imp. A (EP), Amen, Aragest, Aragest 5, Asconale, CBP 1011, Clinofem, Clinovir, Colirest, Curretab, Cycrin, DMPA, Depo-Clinovir, Depo-Prodasone, Depo-Provera, Depo-Ralovera, Depo-SubQ Provera 104, Depo-progestin, Depo-promone, Depomedroxyprogesterone acetate, Depot-medroxyprogesterone acetate, Farlutal, Farlutin, G-Farlutal, Gestapuran, Hematrol, Hysron, Indivina, Lutopolar, Lutoral, MAP, MAP (steroid), MPA, MPA Gyn 5, Med-Pro, Medroprogesterone acetate, Medroxyprogesterone 17-acetate, Meprate, Metigestrona, NSC 21171, NSC 26386, Nadigest, Nidaxin, Novo-Medrone, Oragest, Perlutex, Perlutex Leo, Prempack C, Prodasone, Progespon, Progestalfa, Progeston, Progevera, Provera, Proverone, Ralovera, Repromap, Repromix, Sirprogen, Sodelut G, U 8839, Veramix, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methyl-, (6alpha)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6alpha-methyl-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6alpha-methyl-, acetate (6CI), (6alpha)-17-(Acetyloxy)-6-methylpregn-4-ene-3,20-dione, 17alpha-Acetoxy-6alpha-methylpregn-4-ene-3,20-dione, 17alpha-Acetoxy-6alpha-methylprogesterone, 17alpha-Hydroxy-6alpha-methyl-4-pregnene-3,20-dione 17-acetate, 17alpha-Hydroxy-6alpha-methylpregn-4-ene-3,20-dione acetate, 17alpha-Hydroxy-6alpha-methylprogesterone acetate, 6alpha-Methyl-17-acetoxyprogesterone, 6alpha-Methyl-17R-hydroxyprogesterone acetate, 6alpha-Methyl-17alpha-Hydroxy-4-pregnene-3,20-dione acetate, 6alpha-Methyl-17alpha-acetoxy-Delta4-pregnene-3,20-dione, 6alpha-Methyl-17alpha-acetoxypregn-4-ene-3,20-dione, 6alpha-Methyl-17alpha-hydroxyprogesterone acetate, Megestrol Acetate Impurity A
InChI Formula InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1
SMILES C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C)C(=O)C)[C@@]4(C)CCC(=O)C=C14
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 386.52
Quantity 250 mg
Formula Weight 386.2457 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.