missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medroxyprogesterone Acetate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Medroxyprogesterone acetate |
| CAS | 71-58-9 |
| Analyte or Component Names | Medroxyprogesterone acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H34 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6alpha-Methyl-3,20-dioxopregn-4-en-17-yl acetate, Medroxyprogesterone acetate, Megestrol Acetate Imp. A (EP), Amen, Aragest, Aragest 5, Asconale, CBP 1011, Clinofem, Clinovir, Colirest, Curretab, Cycrin, DMPA, Depo-Clinovir, Depo-Prodasone, Depo-Provera, Depo-Ralovera, Depo-SubQ Provera 104, Depo-progestin, Depo-promone, Depomedroxyprogesterone acetate, Depot-medroxyprogesterone acetate, Farlutal, Farlutin, G-Farlutal, Gestapuran, Hematrol, Hysron, Indivina, Lutopolar, Lutoral, MAP, MAP (steroid), MPA, MPA Gyn 5, Med-Pro, Medroprogesterone acetate, Medroxyprogesterone 17-acetate, Meprate, Metigestrona, NSC 21171, NSC 26386, Nadigest, Nidaxin, Novo-Medrone, Oragest, Perlutex, Perlutex Leo, Prempack C, Prodasone, Progespon, Progestalfa, Progeston, Progevera, Provera, Proverone, Ralovera, Repromap, Repromix, Sirprogen, Sodelut G, U 8839, Veramix, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methyl-, (6alpha)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6alpha-methyl-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6alpha-methyl-, acetate (6CI), (6alpha)-17-(Acetyloxy)-6-methylpregn-4-ene-3,20-dione, 17alpha-Acetoxy-6alpha-methylpregn-4-ene-3,20-dione, 17alpha-Acetoxy-6alpha-methylprogesterone, 17alpha-Hydroxy-6alpha-methyl-4-pregnene-3,20-dione 17-acetate, 17alpha-Hydroxy-6alpha-methylpregn-4-ene-3,20-dione acetate, 17alpha-Hydroxy-6alpha-methylprogesterone acetate, 6alpha-Methyl-17-acetoxyprogesterone, 6alpha-Methyl-17R-hydroxyprogesterone acetate, 6alpha-Methyl-17alpha-Hydroxy-4-pregnene-3,20-dione acetate, 6alpha-Methyl-17alpha-acetoxy-Delta4-pregnene-3,20-dione, 6alpha-Methyl-17alpha-acetoxypregn-4-ene-3,20-dione, 6alpha-Methyl-17alpha-hydroxyprogesterone acetate, Megestrol Acetate Impurity A |
| InChI Formula | InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C)C(=O)C)[C@@]4(C)CCC(=O)C=C14 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction