Learn More
Medical Viscosity Standard, 4.0 mPa.s (25.00°C ), Paragon Scientific™
Product Code. 16000240
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 16000240
Brand: Paragon Scientific MGVS40500
Tested in strict accordance with ASTM D2162. These medical grade viscosity standards are fully traceable to national standards and are also test equipment compatible. Typically used, but not limited to, the calibration and verification of viscosity measuring equipment.
  • Fully traceable to international standards
  • Certified in accordance to ISO 17025 & ISO 17034 under Paragon's UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Tamper evident security packaging
  • 12 month shelf life
  • Manufactured in the United Kingdom
  • Contains antibacterial agent
Specifications
Certifications/Compliance ISO 17025, ISO 17034
Quantity 500 mL
Nominal values quoted. Certified Values will vary from batch to batch.