missing translation for 'onlineSavingsMsg'
Learn More

MED31 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

466.00€

Specifications

Antigen MED31
Applications Western Blot, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18217253
View Documents
Novus Biologicals
NBP1-56861
100 μL
466.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MED31 Polyclonal specifically detects MED31 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications

Specifications

MED31
Polyclonal
Rabbit
Q9Y3C7
51003
Synthetic peptides corresponding to MED31(mediator complex subunit 31) The peptide sequence was selected from the N terminal of MED31. Peptide sequence MAAAVAMETDDAGNRLRFQLELEFVQCLANPNYLNFLAQRGYFKDKAFVN.
Primary
Western Blot, ChIP Assay
Unconjugated
RUO
CGI-125, FLJ27436, FLJ36714, hSOH1, mediator complex subunit 313110004H13Rik, Mediator complex subunit SOH1, mediator of RNA polymerase II transcription subunit 31, mediator of RNA polymerase II transcription, subunit 31 homolog, mediator of RNA polymerase II transcription, subunit 31 homolog (S. cerevisiae), SOH1
MED31
IgG
This product is specific to Subunit or Isoform: 31.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.