missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35228-100ul
This item is not returnable.
View return policy
Description
MED21 Polyclonal antibody specifically detects MED21 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MED21 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| hSrb7, mediator complex subunit 21SRB7 (suppressor of RNA polymerase B, yeast) homolog, RNA polymerase II complex component SRB7, RNA polymerase II holoenzyme component SRB7, RNAPII complex component SRB7, SRB7 suppressor of RNA polymerase B homolog, SRB7SRB7 suppressor of RNA polymerase B homolog (yeast), SURB7mediator of RNA polymerase II transcription subunit 21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human MED21 (NP_004255.2).,, Sequence:, MADRLTQLQDAVNSLADQFCNAIGVLQQCGPPASFNNIQTAINKDQPANPTEEYAQLFAALIARTAKDIDVLIDSLPSEESTAALQAASLYKLEEENHEAATCLEDVVYRGDMLLEKIQSALADIAQSQLKTRSGTHSQSLPDS | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 9412 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction