missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MCT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30936-100ug
This item is not returnable.
View return policy
Description
MCT2 Polyclonal antibody specifically detects MCT2 in Human samples. It is validated for Western Blot
Specifications
| MCT2 | |
| Polyclonal | |
| Western Blot : 1:400 | |
| MCT 2, MCT2solute carrier family 16 (monocarboxylic acid transporters), member 7, monocarboxylate transporter 2, Solute carrier family 16 member 7, solute carrier family 16, member 7 (monocarboxylic acid transporter 2) | |
| Synthetic peptide derived from human MCT2 protein | |
| 100 μg | |
| Lipid and Metabolism | |
| 9194 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction