missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCPIP1/ZC3H12A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€
Specifications
| Antigen | MCPIP1/ZC3H12A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
MCPIP1/ZC3H12A Polyclonal specifically detects MCPIP1/ZC3H12A in Human samples. It is validated for Western Blot, ELISA.Specifications
| MCPIP1/ZC3H12A | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Editing and Processing Endonucleases, Immunology, Inflammation | |
| PBS with 0.02% Sodium Azide | |
| Reg1, Regnase-1, zinc finger CCCH-type containing 12A | |
| ZC3H12A | |
| IgG | |
| Affinity Purified | |
| This ZC3H12A antibody is predicted to have no cross-reactivity to other members of the ZC3H12 protein family. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_079355 | |
| 80149 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human ZC3H12A. The immunogen is located within amino acids 330 - 380 of ZC3H12A. Amino Acid Squence: PPRAPSKDKNGRRPSPS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 6 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title