missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCPH1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148789
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.
Specifications
| MCPH1 | |
| Polyclonal | |
| Unconjugated | |
| MCPH1 | |
| 5430437K10Rik; BRCT-repeat inhibitor of TERT expression 1; BRIT 1; BRIT1; D030046N04Rik; FLJ12847; Mcph1; MCT; Microcephalin; microcephalin 1; microcephaly, primary autosomal recessive 1; RGD1560642; truncated microcephalin | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 79648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8NEM0 | |
| MCPH1 | |
| E. coli-derived recombinant human MCPH1, Met1-Lys250. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction