missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCL1 Monoclonal Antibody (OTI10D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500990S
This item is not returnable.
View return policy
Description
MCL1 Monoclonal antibody specifically detects MCL1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MCL1 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q07820 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4170 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI10D10 | |
| Unconjugated | |
| MCL1 | |
| AW556805; BCL2 family apoptosis regulator; BCL2L3; bcl2-L-3; Bcl-2-like protein 3; bcl-2-related protein EAT/mcl1; EAT; Induced myeloid leukemia cell differentiation protein Mcl-1; induced myeloid leukemia cell differentiation protein Mcl-1 homolog; Mcl1; Mcl-1; mcl1/EAT; MCL1-ES; MCL1L; MCL1S; MCL-1S; MGC104264; MGC1839; myeloid cell leukemia 1; myeloid cell leukemia ES; myeloid cell leukemia sequence 1; myeloid cell leukemia sequence 1 (BCL2-related); TM | |
| Full length human recombinant protein of human MCL1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction