missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mcl-1 Antibody (8C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2194
This item is not returnable.
View return policy
Description
Mcl-1 Monoclonal antibody specifically detects Mcl-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Mcl-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| bcl2-L-3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM | |
| Purified recombinant fragment of human Mcl-1 expressed in E. Coli. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 4170 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8C6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction