missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCHR1 Monoclonal Antibody (1032149)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561888

Product Code. 30534354

  • 419.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MCHR1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
P97639, Q8JZL2, Q99705
Mchr1
E. coli-derived human MCHR1, Met1-Pro113.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
1032149
Unconjugated
Mchr1
AW049955; G protein coupled receptor 24; G protein-coupled receptor 24; GPCR24; GPR24; Gpr24-9 protein coupled receptor; G-protein coupled receptor 24; MCH 1 receptor; MCH receptor 1; MCH1; Mch1r; MCH-1R; MCHR; MCHR1; MCH-R1; MCHR-1; melanin concentrating hormone receptor 1; melanin-concentrating hormone receptor 1; SLC1; SLC-1; Somatostatin receptor-like protein
Mouse
Protein A/G
RUO
207911, 2847, 83567
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.