missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCF2 Monoclonal Antibody (OTI2C12), TrueMAB™, OriGene
Description
MCF2 Monoclonal antibody specifically detects MCF2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antigen | MCF2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | MASP2 |
| Gene Accession No. | O00187 |
| Gene Alias | ARHGEF21; B230117G22Rik; DBL; DBL-transforming protein; MCF.2 cell line derived transforming sequence; mcf.2 transforming sequence; MCF2; Mcf-2; MCF2-transforming protein; Oncogene MCF2 (oncogene DBL); OTTHUMP00000024159; proto-oncogene DBL; Proto-oncogene MCF-2; RGD1566098 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?