missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (c264), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28333
This item is not returnable.
View return policy
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Chicken, Turkey samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunocytochemistry.
Specifications
| MCAM/CD146 | |
| Monoclonal | |
| 0.5mg/mL | |
| Flow Cytometry 1 ug/10 cells, Immunoprecipitation 1:10-1:500, Immunocytochemistry | |
| MCAM | |
| Mixture of E15 and E16 thymocytes from H.B15, H.B19, and H.B21 chicken embryos | |
| 0.25 mg | |
| Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 4162 | |
| Chicken, Turkey | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| c264 | |
| Biotin | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken MCAM, Mr 98 kDa (protein core approx. 84 kDa) | |
| Store at 4C. Do not freeze. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction