missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad50 Antibody (S05-5H6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19512-50ul
This item is not returnable.
View return policy
Description
Rad50 Monoclonal antibody specifically detects Rad50 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Rad50 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S05-5H6 | |
| Unconjugated | |
| DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2 | |
| A synthetic peptide of human Rad50 (Uniprot # Q92878) | |
| 50 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining | |
| 10111 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction