missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MBP Monoclonal antibody specifically detects MBP in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MBP |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL2827 |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P02686, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?