Learn More
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Product Code. p-4526986 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 x 1.25mL
1000U
2 x 1.25mL
500U
Unit Size:
100 reactions
500 reactions
500 units
Pack of 5
Product Code. 10394380
Brand: Thermo Scientific™ EP0602
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
In addition, the enzyme provides the convenience of reaction set-up at room temperature. Maxima Hot Start Taq DNA Polymerase is a recombinant Taq DNA polymerase which has been chemically modified by the addition of heat-labile blocking groups to its amino acid residues. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification (see Fig. 1). The functional activity of the enzyme is restored during a short 4-minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase: catalyzes 5'=>3' synthesis of DNA, has no detectable 3'=>5' proofreading exonuclease activity, but possesses low 5'=>3' exonuclease activity. It exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Before activation, the two activities are not detectable.
  • High yield amplification of complex templates
  • Four minutes activation time
  • High PCR specificit—reduced effects of mispriming and primer-dimer formation
  • Enhanced PCR sensitivity
  • Convenient room temperature PCR set-up
  • Generates PCR products with 3'-dA overhangs
Recommended for:
Hot start PCR; High yield amplification of targets up to 3kb; RT-PCR.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2 x 1.25mL of 10X Hot Start PCR Buffer, 2 x 1.25mL of 25mM MgCl2
Description Maxima™ Hot Start™ Taq DNA Polymerase
Quantity 500U
For Use With (Application) Cloning