Learn More
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Product Code. 10763721 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1.25mL
1000U
2 x 1.25mL
Unit Size:
100 reactions
500 reactions
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10763721 1000U Pack of 5
11886833 2 x 1.25mL 100 reactions
11883923 10 x 1.25mL 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10763721 Supplier Thermo Scientific™ Supplier No. EP0603
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
Highlights
  • Four minute activation time
  • High PCR specificity and sensitivity
  • Room temperature PCR set up
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 10 x 1.25mL of 10X Hot Start PCR Buffer, 10 x 1.25mL of 25mM MgCl2
Description Maxima™ Hot Start™ Taq DNA Polymerase
Quantity 1000U
For Use With (Application) Cloning