missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Massachusetts Residual Solvents - FET Mixture 243 1000 μg/mL in Triacetin, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 67-64-1, 75-05-8, 64-17-5, 75-28-5, 67-63-0, 67-56-1 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Triacetin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction