Learn More
IBA Lifesciences MART-1 peptide ELAGIGILTV (HLA-A*0201)
GREENER_CHOICE
Product Code. 15322126 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 15322126
Brand: IBA Lifesciences 67007901
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • MART-1 peptide ELAGIGILTV (HLA-A*0201) is a single peptide for stimulation of T cells.
  • The peptide from melanoma antigen recognized by T cells 1 (MART-1) is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
  • Specifications
    Content And Storage Recommended: -25°C to -15°C
    Quantity 1 mg