missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Marmoset OX40 / TNFRSF4 / CD134 Protein, His Tag
535.63€ - 3919.27€
Specifications
Accession Number | XP_002750223.1 |
---|---|
Molecular Weight (g/mol) | 21.8 kDa |
Source | Marmoset O x 40, His Tag (O x 0-M522b) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 214 (Accession # F7DHA3-1). |
Immunogen | Leu 29 - Ala 214 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16497929
|
ACROBiosystems
OX0-M522B-1MG |
1 mg |
3919.27€
1mg |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
15817407
|
ACROBiosystems
OX0-M522B-100UG |
100 μg |
535.63€
100µg |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.8 kDa. The protein migrates as 42-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
XP_002750223.1 | |
Marmoset O x 40, His Tag (O x 0-M522b) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 214 (Accession # F7DHA3-1). | |
-20°C | |
OX40 | |
Marmoset | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
21.8 kDa | |
Leu 29 - Ala 214 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |