Learn More
ACROBiosystems Marmoset OX40 / TNFRSF4 / CD134 Protein, His Tag
Brand: ACROBiosystems OX0-M522b-1mg
Additional Details : Weight : 0.00010kg
Description
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.8 kDa. The protein migrates as 42-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
XP_002750223.1 | |
1 mg | |
Leu 29 - Ala 214 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
21.8 kDa | |
Marmoset O x 40, His Tag (O x 0-M522b) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 214 (Accession # F7DHA3-1). | |
-20°C | |
OX40 | |
Marmoset | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |