missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK2 Antibody (S07-1F5), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Specifications
| Antigen | MAPKAPK2 |
|---|---|
| Clone | S07-1F5 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686514
|
Novus Biologicals
NBP3-19891-50ul |
50 μg |
421.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671715
|
Novus Biologicals
NBP3-19891-100ul |
100 μg |
645.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAPKAPK2 Monoclonal antibody specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MAPKAPK2 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 | |
| A synthetic peptide of human MAPKAPK2 (Uniprot # P49137) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-1F5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 9261 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title