missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (CL5420) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61417V
This item is not returnable.
View return policy
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Single Cell Western,Immunocytochemistry/ Immunofluorescence
Specifications
| MAP2 | |
| Monoclonal | |
| DyLight 405 | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| This antibody was developed using a recombinant protein derived from P11137, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Single Cell Western, Immunocytochemistry/Immunofluorescence | |
| CL5420 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction