Learn More
MAN1A2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14049-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in the maturation of Asn-linked oligosaccharides. Progressively trim alpha-1, 2-linked mannose residues from Man(9)GlcNAc(2) to produce Man(5)GlcNAc(2).Specifications
| MAN1A2 | |
| Polyclonal | |
| Unconjugated | |
| MAN1A2 | |
| Alpha 1,2 mannosidase IB, MAN1A2, MAN1B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10905 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60476 | |
| MAN1A2 | |
| MAN1A2 Fusion Protein Ag5240 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |