Learn More
IBA Lifesciences pCSG-IBA Mammalian Expression Vector with No Secretion
GREENER_CHOICE
Product Code. 15613868 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pCSG-IBA103
pCSG-IBA105
pCSG-IBA123
pCSG-IBA143
pCSG-IBA145
pCSG-IBA162
pCSG-IBA164
pCSG-IBA167
pCSG-IBA168
pCSG-IBA23
pCSG-IBA3
pCSG-IBA43
pCSG-IBA45
pCSG-IBA5
pCSG-IBA62
pCSG-IBA64
pCSG-IBAwt1
Unit Size:
5µg
Product Code. 15613868
Brand: IBA Lifesciences 55064001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for high-level episomal expression in mammalian hosts under G418 selection. IBA Lifesciences GmbH Mammalian Expression Vector with No Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).
pCSG-IBA mammalian expression vectors
  • Designed for high-level episomal expression in mammalian hosts under G418 selection containing the following elements:
    • Human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells
    • Episomal replication through Epstein Barr Virus replication origin (oriP) and nuclear antigen encoded by EBNA-1 in human, primate and canine cells and through SV40 replication origin in cells latently infected with SV40 or that express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7)
  • Stable cell lines can be selected by the neomycin resistance gene (NeoR).
  • In addition, the human cytomegalovirus (CMV) immediate-early promoter enables a high-level expression in a wide range of mammalian cells.
Please note that cloning into pCSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. Besides to the direct cloning of the gene of interest into pCSG-IBA vectors with Esp3I, another option via a so-called entry-vector is possible.
Specifications
Constitutive or Inducible System Constitutive
Marker Gene Ampicillin
Promoter CMV
Product Type Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Species Mammalian
Protein Tag Strep-tag II, FLAG-tag
Cloning Method Direct Cloning
Concentration 250 ng/μL
Primer Forward
Quantity 5 μg
Vector pCSG-IBA64
For Use With (Application) Episomal expression (oriP, EBNA-1)