missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Mouse anti-Human, Clone: MGB/4056, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07423-100ug
This item is not returnable.
View return policy
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Mammaglobin A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| Recombinant full-length human Mammaglobin A protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Peptide Array | |
| MGB/4056 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4250 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction