missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (3C8), Novus Biologicals™
Mouse Monoclonal Antibody
215.00€
Specifications
| Antigen | Mammaglobin A |
|---|---|
| Clone | 3C8 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Mammaglobin A | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| SCGB2A2 | |
| IgG1 | |
| Unpurified | |
| 11 kDa |
| 3C8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 4250 | |
| Purified recombinant fragment of human SCGB2A2 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title