missing translation for 'onlineSavingsMsg'
Learn More

MAML3 Antibody (MAML3/1303), DyLight 405, Novus Biologicals™

Product Code. 18745621 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18745621 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18745621 Supplier Novus Biologicals Supplier No. NBP254309V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAML3 Monoclonal antibody specifically detects MAML3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAML3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAML3/1303
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias CAG/CTG 3, ERDA3, MAM-2, mam-3, mastermind-like 3 (Drosophila), mastermind-like protein 3, polyglutamine rich, trinucleotide repeat containing 3
Host Species Mouse
Immunogen Recombinant human MAML3 protein fragment (Uniprot: Q96JK9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 55534
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.