missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin 1, Mouse anti-Human, Clone: rVIL1/1325, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79809-20UG
This item is not returnable.
View return policy
Description
Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Villin 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P09327 | |
| VIL1 | |
| A recombinant fragment (around aa179-311) of human Villin 1 protein (exact sequence is proprietary) | |
| 20 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 7429 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| rVIL1/1325 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| D2S1471, VIL, villin 1, villin-1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody recognizes a protein of 95kDa, which is identified as Villin 1. It is a major constituent in the microvilli, which compose the brush border of epithelial cells forming absorptive surfaces of the intestinal and renal proximal tubular epithelia. Anti-Villin 1 labels the brush border area in the gastrointestinal mucosal epithelium and urogenital tract. Among neoplasms, villin is predominantly expressed in tumors of colorectal origin. This antibody is useful in identifying malignant cells from primary and metastatic colorectal carcinomas; it also labels Merkel cells of the skin. | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction