Learn More
IBA Lifesciences MagStrep 'Type3' XT Beads
Product Code. 16554594 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16554594 10 mL 10mL
15362006 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16554594 Supplier IBA Lifesciences Supplier No. 24090010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
MagStrep 'type3' XT beads are magnetic beads (5% suspension) to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • MagStrep “type3” XT beads are magnetic beads consisting of a 6% agarose coated with the streptavidin variant Strep-Tactin™XT.
  • Due to the magnetic core no centrifugation step is required since the beads can be easily and swiftly separated with a magnetic separator, reducing the risk of disturbing the pellet or loss of beads within the wash step to zero.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • MagStrep “type3” XT beads provide the highest binding capacity.
  • These characteristics make them a perfect tool for isolation of proteins or complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP) in batch format and/or high-throughput screenings.
  • The elution of target proteins or complexes can be triggered under mild conditions with the biotin-containing Buffer BXT or under denaturing conditions by boiling in SDS gel loading buffer.
  • 20 μL suspension correspond to 1 μL magnetic beads.
Specifications
Content And Storage 2°C to 8°C. Do not freeze.
Product Type Strep-Tactin Coated Magnetic Beads
Format Beads
Quantity 10 mL
Sample Type 3
For Research Use Only.