missing translation for 'onlineSavingsMsg'
Learn More

Abcam Magnesium Dodecyl Sulfate, Anionic Detergent, Solid, 25 g

Product Code. 32108430
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32108430 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32108430 Supplier Abcam Supplier No. ab14629725g
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High purity anionic detergent for use in chemical research.

  • Molecular weight: 551.1 g/mol
  • Purity: >98%
  • Form: Solid
  • Product type: Anionic detergent
  • Composition: 98% Magnesium dodecyl sulfate

Specifications

Buffer 98% Magnesium dodecyl sulfate
Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Formulation 98% Magnesium dodecyl sulfate
Quantity 25 g
For Use With (Application) Sample preparation and detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.