missing translation for 'onlineSavingsMsg'
Learn More

MAGED4B Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30566567 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30566567 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30566567 Supplier Novus Biologicals Supplier No. NBP345970MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAGED4B Polyclonal antibody specifically detects MAGED4B in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGED4B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias KIAA1859FLJ39060, MAGE1, MAGE-D4 antigen, MAGE-E1, MAGE-E1 antigen, MAGEE1MAGED4A, melanoma antigen family D, 4, MGC3210
Host Species Rabbit
Immunogen melanoma antigen family D, 4. (Uniprot# Q96JG8)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81557
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.