missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Antibody (21B4) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
267.00€ - 640.00€
Specifications
| Antigen | MAGEA3 |
|---|---|
| Clone | 21B4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650835
|
Novus Biologicals
NBP3-20167-0.025mg |
0.025 mg |
267.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661675
|
Novus Biologicals
NBP3-20167-0.2mg |
0.2 mg |
640.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| MAGEA3 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 | |
| The original antibody was isolated from a patient suffering from a tumor expressing a tumor specific antigen like MAGE-A3. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 21B4 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS | |
| 4102 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title