missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MafB Antibody (OTI1E9), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72569IR
This item is not returnable.
View return policy
Description
MafB Monoclonal specifically detects MafB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| MafB | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAFB | |
| Recombinant protein fragment corresponding to amino acids 32-320 of human MafB (NP_005452) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1E9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Kreisler (mouse) maf-related leucine zipper homolog, Kreisler maf-related leucine zipper homolog, KRMLMAFB/Kreisler basic region/leucine zipper transcription factor, Maf-B, MGC43127, transcription factor MafB, V-maf musculoaponeurotic fibrosarcoma oncogene homolog B, v-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian) | |
| Mouse | |
| Affinity Purified | |
| Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 9935.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction