missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macro H2A.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 307.65€
Specifications
| Antigen | Macro H2A.2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637132
|
Novus Biologicals
NBP2-97224-100ul |
100 μg |
326.00€ 307.65€ / 100µL Save 18.35€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672502
|
Novus Biologicals
NBP2-97224-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Macro H2A.2 Polyclonal antibody specifically detects Macro H2A.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Macro H2A.2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
| Produced in rabbits immunized with E. coli-derived Human Macro H2A.2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 55506 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit