missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MACC1 Antibody (CL0856) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52955AF647
This item is not returnable.
View return policy
Description
MACC1 Monoclonal antibody specifically detects MACC1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MACC1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| metastasis associated in colon cancer 1, SH3 domain-containing protein 7a5,7A5 | |
| This antibody was developed using a recombinant protein derived from Q6ZN28, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 346389 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0856 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction