Learn More
Applied Biosystems™ M13 Sequencing Forward Primer, HPLC
Product Code. 15554374 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15554374 50 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15554374 Supplier Applied Biosystems™ Supplier No. A15637
 Restricted Item
Estimated Shipment: 02-12-2026
M13 Sequencing Forward Primer, HPLC
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY
Specifications
Delivery Type Single Tube, Dried-down
Format Solid
For Use With (Equipment) Mass Spectrometry
Description M13 (-21) Forward Sequencing Primer, 50 nmole, HPLC Purification, 900pmole Minimal Spec Yield, 5' TGTAAAACGACGGCCAGT Sequence, Delivered in a Single tube, Dried-down, Used for BigDye™ Cycle Sequencing Primers
Product Type M13 (-21) Forward Sequencing Primer
Protein Degradation Sequence 5' TGTAAAACGACGGCCAGT
Primer M13 (-21) Forward Sequencing
Quantity 50 nmol
Grade HPLC
For Use With (Application) BigDye™ Cycle Sequencing Primers
Form Solid