missing translation for 'onlineSavingsMsg'
Learn More

M-Benzyl-N-trifluoroacetyl-L-tryptophan Methyl Ester, TRC

Product Code. 30332320
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
This item is not returnable. View return policy

Product Code. 30332320

Brand: Toronto Research Chemicals TRCB316145250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material M-Benzyl-N-trifluoroacetyl-L-tryptophan Methyl Ester
Analyte or Component Names M-Benzyl-N-trifluoroacetyl-L-tryptophan Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C21 H19 F3 N2 O3
InChI Formula InChI=1S/C21H19F3N2O3/c1-29-19(27)18(11-15-12-25-17-10-6-5-9-16(15)17)26(20(28)21(22,23)24)13-14-7-3-2-4-8-14/h2-10,12,18,25H,11,13H2,1H3/t18-/m0/s1
Solution Type Neat
SMILES COC(=O)[C@H](Cc1c[nH]c2ccccc12)N(Cc3ccccc3)C(=O)C(F)(F)F
IUPAC Name methyl (2S)-2-[benzyl-(2,2,2-trifluoroacetyl)amino]-3-(1H-indol-3-yl)propanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 404.382
Quantity 250 mg
Formula Weight 404.135
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.