missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41152
This item is not returnable.
View return policy
Description
LZTS2 Polyclonal specifically detects LZTS2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| LZTS2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LZTS2 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human LZTS2. The immunogen is located within amino acids 80 - 130 of LZTS2. Amino Acid Squence: INEDFRTESPPSPSSDVED | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 84445 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, ELISA, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml | |
| hLZTS2, KIAA1813leucine zipper putative tumor suppressor 2, LAPSER1Protein LAPSER1, leucine zipper, putative tumor suppressor 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| LZTS2 antibody is predicted to not cross-react with other LZTS family members | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction