missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTFL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97499-50ul
This item is not returnable.
View return policy
Description
LZTFL1 Polyclonal antibody specifically detects LZTFL1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| LZTFL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| FLJ36386, leucine zipper transcription factor-like 1, leucine zipper transcription factor-like protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human LZTFL1 fragment. | |
| 50 μg | |
| Epigenetics | |
| 54585 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction