Learn More
LYVE-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 19 publications
Brand:  Novus Biologicals NB600-1008SS
 View more versions of this product
Product Code. 18011384
  • 197.00€ / 25µg
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LYVE-1
Polyclonal
Unconjugated
PBS with No Preservative
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1
Rabbit
35 kDa
0.025 mg
Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis
10894
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunofluorescence, CyTOF
LYOPH
Western Blot 1-2 ug/mL, Flow Cytometry 3-20 ug/mL, ELISA 1-15 ug/mL, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:800, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating, Immunofluorescence, CyTOF-ready
Q8BHC0
LYVE1
Produced from sera of rabbits immunized with highly pure recombinant mouse soluble LYVE-1 produced in insect cells. The recombinant soluble LYVE-1 consists of amino acid 24 (Ala) to 228 (Gly) and is fused to a C-terminal His-tag (6xHis).
Protein A purified
RUO
Primary
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. Please note the sample size of this product will be provided in reconstituted liquid form.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.