missing translation for 'onlineSavingsMsg'
Learn More

Lysozyme Antibody, Biotin, Novus Biologicals™

Product Code. 18062596 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.002 g
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062596 0.002 g 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18062596 Supplier Novus Biologicals Supplier No. NB10020050.002G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lysozyme Polyclonal specifically detects Lysozyme in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysozyme
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:2000-1:10000, ELISA 1:20000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Gene Alias EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Gene Symbols LYZ
Host Species Rabbit
Immunogen This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length protein corresponding to amino acids 1-129 of Hen Egg White Lysozyme.
Quantity 0.002 g
Gene ID (Entrez) 4069
Test Specificity This purified antibody is directed against lysozyme from hen egg white protein.
Target Species Chicken
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Marker Histiocytic Marker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.