missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody (7H3D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€
Specifications
| Antigen | Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B |
|---|---|
| Clone | 7H3D7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Monoclonal specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Modifiers, Epigenetics | |
| PBS with 0.05% Sodium Azide | |
| 10765 | |
| Purified recombinant fragment of human KDM5B (AA: 231-319) expressed in E. Coli. | |
| Primary | |
| 175.6 kDa |
| 7H3D7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
| KDM5B | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title